Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-7-chloroimidazo[1,2-c]pyrimidine-2-carboxylate ≥95%

Supplier: MATRIX SCIENTIFIC MS

070249-1G 070249-5G
101943-952EA 1687.69 USD InStock
101943-952 101943-954
Ethyl-7-chloroimidazo[1,2-c]pyrimidine-2-carboxylate ≥95%
Ethyl-7-chloroimidazo[1,2-c]pyrimidine-2-carboxylate
Hazard: Irritant


Formula: C₉H₈ClN₃O₂
MDL Number: MFCD11045730
CAS Number: 1105187-40-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty