070276-1G
070276-5G
101944-060EA
1832.31
USD
InStock
101944-060
101944-062
4,5,6,7-Tetrahydro-1H-pyrazolo[3,4-c]pyridine hydrochloride ≥95%
4,5,6,7-Tetrahydro-1H-pyrazolo[3,4-c]pyridine hydrochloride
Purity: 95%
CAS: 879668-17-4
MDL Number: MFCD11846200
Molecular Formula: C6H10ClN3
Molecular Weight: 159,62
Hazard: Irritant