Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,5,6,7-Tetrahydro-2H-pyrazolo[4,3-b]pyridine hydrochloride ≥95%

Supplier: MATRIX SCIENTIFIC MS

070277-1G 070277-5G
101944-064EA 2093.85 USD InStock
101944-064 101944-066
4,5,6,7-Tetrahydro-2H-pyrazolo[4,3-b]pyridine hydrochloride ≥95%
4,5,6,7-Tetrahydro-2H-pyrazolo[4,3-b]pyridine hydrochloride
Hazard: Irritant


Formula: C₆H₁₀ClN₃
MDL Number: MFCD11846201
CAS Number: 1187830-47-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty