Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-Tyr(tBu)-Bt ≥98%

Supplier: MATRIX SCIENTIFIC MS

070304-250MG 070304-500MG
101944-172EA 1106.15 USD InStock
101944-172 101944-174
Fmoc-Tyr(tBu)-Bt ≥98%
Fmoc-Tyr(tBu)-Bt
Hazard: Irritant


Formula: C₃₄H₃₂N₄O₄
MDL Number: MFCD11850165
CAS Number: 1126433-42-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty