Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Cyclopropylmethoxy-4,5-difluorophenylboronicacid pinacol ester ≥96%

Supplier: MATRIX SCIENTIFIC MS

070329-500MG 070329-1G
101944-280EA 524.62 USD InStock
101944-280 101944-282
2-Cyclopropylmethoxy-4,5-difluorophenylboronicacid pinacol ester ≥96%
2-Cyclopropylmethoxy-4,5-difluorophenylboronicacid pinacol ester
Hazard: Irritant


Formula: C₁₆H₂₁BF₂O₃
MW: 310.15 g/mol
MDL Number: MFCD14702521
CAS Number: 1245816-08-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty