Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Benzyloxy-5-methylphenylboronic acid pinacol ester ≥98%

Supplier: MATRIX SCIENTIFIC MS

070330-500MG 070330-1G
101944-284EA 524.62 USD InStock
101944-284 101944-286
2-Benzyloxy-5-methylphenylboronic acid pinacol ester ≥98%
2-Benzyloxy-5-methylphenylboronic acid pinacol ester
Hazard: Irritant


Formula: C₂₀H₂₅BO₃
MDL Number: MFCD13152011
CAS Number: 1204580-85-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty