Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5H-Imidazo[1,2-b]pyrazole-2-carboxylic acid ≥98%

Supplier: MATRIX SCIENTIFIC MS

070373-1G 070373-5G
101944-456EA 1823.08 USD InStock
101944-456 101944-458
5H-Imidazo[1,2-b]pyrazole-2-carboxylic acid ≥98%
5H-Imidazo[1,2-b]pyrazole-2-carboxylic acid
Hazard: Irritant


Formula: C₆H₅N₃O₂
MDL Number: MFCD08056315
CAS Number: 914637-58-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty