Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(E)-1-Ethoxyethene-2-boronic acid pinacolester ≥97%

Supplier: MATRIX SCIENTIFIC MS

070394-1G 070394-5G
101944-556EA 209.23 USD InStock
101944-556 101944-558
(E)-1-Ethoxyethene-2-boronic acid pinacolester ≥97%
(E)-1-Ethoxyethene-2-boronic acid pinacolester
Hazard: Irritant/Store in Freezer


Formula: C₁₀H₁₉BO₃
MDL Number: MFCD09998813
CAS Number: 1201905-61-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty