Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl perfluoro-1,10-decanedicarboxylate ≥96%

Supplier: MATRIX SCIENTIFIC MS

005680-5G 005680-25G 005680-100G
101789-332EA 46.15 USD InStock
101789-332 101789-334 101789-336
Dimethyl perfluoro-1,10-decanedicarboxylate ≥96%
Dimethyl perfluoro-1,10-decanedicarboxylate
Hazard: Irritant


Formula: C₁₄H₆F₂₀O₄
MW: 618.17 g/mol
Melting Pt: 33…40 °C
MDL Number: MFCD00236606
CAS Number: 84750-88-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty