Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl tridecafluoroheptanoate ≥98%

Supplier: MATRIX SCIENTIFIC MS

005781-5G 005781-25G
101789-502EA 43.08 USD InStock
101789-502 101789-504
Methyl tridecafluoroheptanoate ≥98%
Methyl tridecafluoroheptanoate
Hazard: Irritant


Formula: C₈H₃F₁₃O₂
Boiling Pt: 138…139 °C
MDL Number: MFCD00153212
CAS Number: 14312-89-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty