Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5,7-Bis(trifluoromethyl)-2-(methylthio)pyrazolo[1,5-a]pyrimidine-3-carboxamide

Supplier: MATRIX SCIENTIFIC MS

005848-250MG 005848-1G
101789-644EA 41.54 USD InStock
101789-644 101789-646
5,7-Bis(trifluoromethyl)-2-(methylthio)pyrazolo[1,5-a]pyrimidine-3-carboxamide
5,7-Bis(trifluoromethyl)-2-(methylthio)pyrazolo[1,5-a]pyrimidine-3-carboxamide
Hazard: Irritant


Formula: C₁₀H₆F₆N₄OS
MW: 344.24 g/mol
MDL Number: MFCD00174394
CAS Number: 175203-36-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty