Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl tetradecafluoroazelate ≥90%

Supplier: MATRIX SCIENTIFIC MS

006280-5G 006280-25G
101790-322EA 69.23 USD InStock
101790-322 101790-324
Dimethyl tetradecafluoroazelate ≥90%
Dimethyl tetradecafluoroazelate
Hazard: Irritant


Formula: C₁₁H₆F₁₄O₄
MDL Number: MFCD00236607
CAS Number: 22116-90-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty