Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Boc-2-pyrroleboronic acid ≥97%

Supplier: MATRIX SCIENTIFIC MS

006553-5G 006553-25G
101790-726EA 44.62 USD InStock
101790-726 101790-728
N-Boc-2-pyrroleboronic acid ≥97%
N-Boc-2-pyrroleboronic acid
Hazard: Refrigerate


Formula: C₉H₁₄BNO₄
Melting Pt: 105…108 °C
MDL Number: MFCD01318939
CAS Number: 135884-31-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty