Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

o-Tolylhydrazinium chloride

Supplier: MATRIX SCIENTIFIC MS

016124-25G 016124-100G
101804-148EA 93.85 USD InStock
101804-148 101804-150
o-Tolylhydrazinium chloride
o-Tolylhydrazinium chloride
Hazard: Irritant


Formula: C₇H₁₁ClN₂
MDL Number: MFCD00012925
CAS Number: 635-26-7
UN: 2811
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty