Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pyrrolidine-1-sulfonyl chloride

Supplier: MATRIX SCIENTIFIC MS

020448-1G 020448-5G 020448-25G
101810-034EA 43.08 USD InStock
101810-034 101810-036 101810-038
Pyrrolidine-1-sulfonyl chloride
Pyrrolidine-1-sulfonyl chloride
Hazard: Irritant


Formula: C₄H₈ClNO₂S
MW: 169.63 g/mol
MDL Number: MFCD07801104
CAS Number: 1689-02-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty