Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-1-fluoro-2-iodobenzene ≥98%

Supplier: MATRIX SCIENTIFIC MS

020562-25G 020562-100G
101810-282EA 29.23 USD InStock
101810-282 101810-284
4-Bromo-1-fluoro-2-iodobenzene ≥98%
4-Bromo-1-fluoro-2-iodobenzene
Hazard: Irritant/Light Sensitive/Refrigerate


Formula: C₆H₃BrFI
MW: 300.9000 g/mol
Boiling Pt: 247 °C
Density: 2.281
Flash Pt: 103 °C
MDL Number: MFCD07368751
CAS Number: 116272-41-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty