Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methyl-4-pyridineboronic acid

Supplier: MATRIX SCIENTIFIC MS

020595-1G 020595-5G
101810-394EA 104.62 USD InStock
101810-394 101810-396
2-Methyl-4-pyridineboronic acid
2-Methyl-4-pyridineboronic acid
Hazard: Irritant/Store in Freezer


Formula: C₆H₈BNO₂
Melting Pt: 122-123°C
MDL Number: MFCD03095364
CAS Number: 579476-63-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty