Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,5,6,7-Tetrahydrothieno[3,2-c]pyridine hydrochloride ≥97%

Supplier: MATRIX SCIENTIFIC MS

027105-25G 027105-100G
101823-504EA 38.46 USD InStock
101823-504 101823-506
4,5,6,7-Tetrahydrothieno[3,2-c]pyridine hydrochloride ≥97%
4,5,6,7-Tetrahydrothieno[3,2-c]pyridine hydrochloride
Hazard: Irritant


Formula: C₇H₁₀ClNS
Melting Pt: 222…224 °C
MDL Number: MFCD05861485
CAS Number: 28783-41-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty