Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Piperidin-4-ylmorpholine

Supplier: MATRIX SCIENTIFIC MS

027114-5G 027114-25G
101823-524EA 53.85 USD InStock
101823-524 101823-526
4-Piperidin-4-ylmorpholine
4-Piperidin-4-ylmorpholine
Hazard: Irritant/Air Sensitive


Formula: C₉H₁₈N₂O
Melting Pt: 40…43 °C
MDL Number: MFCD03274733
CAS Number: 53617-35-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty