Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-o-toluic acid ≥98%

Supplier: MATRIX SCIENTIFIC MS

032352-25G 032352-100G
101833-542EA 44.62 USD InStock
101833-542 102551-146
5-Bromo-o-toluic acid ≥98%
5-Bromo-o-toluic acid
Hazard: Irritant/Store in Freezer


Formula: C₈H₇BrO₂
Melting Pt: 167-171°C
MDL Number: MFCD00267350
CAS Number: 79669-49-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty