Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Bis(trifluoromethyl)fluorobenzene ≥97%

Supplier: MATRIX SCIENTIFIC MS

034721-1G 034721-5G
101838-222EA 43.08 USD InStock
101838-222 101838-224
3,5-Bis(trifluoromethyl)fluorobenzene ≥97%
3,5-Bis(trifluoromethyl)fluorobenzene
Hazard: Irritant/Refrigerate


Formula: C₈H₃F₇
MW: 232.1 g/mol
Boiling Pt: 72…74 °C
MDL Number: MFCD00017997
CAS Number: 35564-19-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty