Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Aminophenylboronic acid hemisulfate

Supplier: MATRIX SCIENTIFIC MS

035492-5G 035492-25G
101839-464EA 186.15 USD InStock
101839-464 101839-466
3-Aminophenylboronic acid hemisulfate
3-Aminophenylboronic acid hemisulphate
Hazard: Irritant


Formula: C₁₂H₁₈B₂N₂O₈S
MDL Number: MFCD00013111
CAS Number: 66472-86-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty