Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Ethoxycarbonyl)phenylboronic acid

Supplier: MATRIX SCIENTIFIC MS

035591-5G 035591-25G
101839-536EA 35.38 USD InStock
101839-536 101839-538
4-(Ethoxycarbonyl)phenylboronic acid
4-(Ethoxycarbonyl)phenylboronic acid
Hazard: Irritant/Store in Freezer


Formula: C₉H₁₁BO₄
MW: 193.99 g/mol
MDL Number: MFCD02179441
CAS Number: 4334-88-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty