Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-6-chloro-5-pivalamidopicolinate

Supplier: MATRIX SCIENTIFIC MS

035723-100MG 035723-500MG
101839-820EA 1073.85 USD InStock
101839-820 101839-822
Methyl-6-chloro-5-pivalamidopicolinate
Methyl-6-chloro-5-pivalamidopicolinate
Hazard: Irritant


Formula: C₁₂H₁₅ClN₂O₃
MDL Number: MFCD11857724
CAS Number: 1142191-95-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty