Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-5-bromo-3-methoxypicolinate

Supplier: MATRIX SCIENTIFIC MS

035767-100MG 035767-500MG
101839-996EA 192.31 USD InStock
101839-996 101839-998
Methyl-5-bromo-3-methoxypicolinate
Methyl 5-bromo-3-methoxypicolinate
Hazard: Irritant


Formula: C₈H₈BrNO₃
MDL Number: MFCD12026771
CAS Number: 1142192-55-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty