Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Furo[3,2-b]pyridin-2-ylmethanol

Supplier: MATRIX SCIENTIFIC MS

035773-100MG 035773-500MG
101840-020EA 900 USD InStock
101840-020 101840-022
Furo[3,2-b]pyridin-2-ylmethanol
Furo[3,2-b]pyridin-2-ylmethanol
Hazard: Irritant


Formula: C₈H₇NO₂
MDL Number: MFCD11857747
CAS Number: 162537-61-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty