Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(1-Methylpiperidin-4-yl)piperazine

Supplier: MATRIX SCIENTIFIC MS

037105-1G 037105-5G 037105-25G
101842-684EA 89.23 USD InStock
101842-684 101842-686 101842-688
1-(1-Methylpiperidin-4-yl)piperazine
1-(1-Methylpiperidin-4-yl)piperazine
Hazard: Irritant


Formula: C₁₀H₂₁N₃
MDL Number: MFCD01075186
CAS Number: 23995-88-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty