Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-(S)-3-amino-4-(3,4-difluorophenyl)butyric acid

Supplier: MATRIX SCIENTIFIC MS

042246-250MG 042246-1G
101855-050EA 373.85 USD InStock
101855-050 101855-052
Boc-(S)-3-amino-4-(3,4-difluorophenyl)butyric acid
Boc-(S)-3-amino-4-(3,4-difluorophenyl)butyric acid
Hazard: Irritant


Formula: C₁₅H₁₉F₂NO₄
MDL Number: MFCD01861050
CAS Number: 270063-54-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty