Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Boc-1,2,5,6-tetrahydropyridine-3-carboxylic acid

Supplier: MATRIX SCIENTIFIC MS

042269-100MG 042269-500MG
101855-142EA 420 USD InStock
101855-142 101855-144
1-Boc-1,2,5,6-tetrahydropyridine-3-carboxylic acid
1-Boc-1,2,5,6-tetrahydropyridine-3-carboxylic acid
Hazard: Irritant


Formula: C₁₁H₁₇NO₄
MDL Number: MFCD11519232
CAS Number: 86447-11-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty