Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-b-Alanine

Supplier: MATRIX SCIENTIFIC MS

042277-10G 042277-25G
101855-170EA 113.85 USD InStock
101855-170 101855-172
Boc-b-Alanine
Boc-b-Alanine
Hazard: Irritant


Formula: C₈H₁₅NO₄
MDL Number: MFCD00037291
CAS Number: 3303-84-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty