Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-(R)-3-amino-4-(3-pyridyl)butyric acid

Supplier: MATRIX SCIENTIFIC MS

042314-250MG 042314-1G
101855-318EA 510.77 USD InStock
101855-318 101855-320
Fmoc-(R)-3-amino-4-(3-pyridyl)butyric acid
Fmoc-(R)-3-amino-4-(3-pyridyl)butyric acid
Hazard: Irritant


Formula: C₂₄H₂₂N₂O₄
MDL Number: MFCD01860959
CAS Number: 269396-66-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty