Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-L-β-homotryptophan

Supplier: MATRIX SCIENTIFIC MS

042372-250MG 042372-1G
101855-544EA 190.77 USD InStock
101855-544 101855-546
Fmoc-L-β-homotryptophan
Fmoc-L-beta-homotryptophan
Hazard: Irritant


Formula: C₂₇H₂₄N₂O₄
MDL Number: MFCD01863056
CAS Number: 353245-98-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty