Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-Nα-methyl-O-benzyl-L-serinedicyclohexylammonium salt

Supplier: MATRIX SCIENTIFIC MS

042449-250MG 042449-1G
101855-848EA 238.46 USD InStock
101855-848 101855-850
Boc-Nα-methyl-O-benzyl-L-serinedicyclohexylammonium salt
Boc-Nalpha-methyl-O-benzyl-L-serinedicyclohexylammonium salt
Hazard: Irritant


Formula: C₂₈H₄₆N₂O₅
MW: 490.69 g/mol
MDL Number: MFCD06795481
CAS Number: 137238-90-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty