Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-2-cyano-3-[4-(trifluoromethyl)phenyl]-acrylate ≥95%

Supplier: MATRIX SCIENTIFIC MS

042716-500MG 042716-1G 042716-5G
101857-158EA 101.54 USD InStock
101857-158 101857-160 101857-162
Ethyl-2-cyano-3-[4-(trifluoromethyl)phenyl]-acrylate ≥95%
Ethyl-2-cyano-3-[4-(trifluoromethyl)phenyl]-acrylate
Hazard: Irritant


Formula: C₁₃H₁₀F₃NO₂
MDL Number: MFCD00176452
CAS Number: 149550-21-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty