Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium-4-iodobenzenesulfonate ≥95%

Supplier: MATRIX SCIENTIFIC MS

042735-500MG 042735-1G 042735-5G
101857-252EA 146.15 USD InStock
101857-252 101857-254 101857-256
Potassium-4-iodobenzenesulfonate ≥95%
Potassium-4-iodobenzenesulfonate
Hazard: Irritant


Formula: C₆H₄IKO₃S
MDL Number: MFCD06656232
CAS Number: 166902-23-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty