Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Boc-3-hydroxymethylindole ≥95%

Supplier: MATRIX SCIENTIFIC MS

043167-500MG 043167-1G 043167-5G
101859-474EA 156.92 USD InStock
101859-474 101859-476 101859-478
1-Boc-3-hydroxymethylindole ≥95%
1-Boc-3-hydroxymethylindole
Hazard: Irritant


Formula: C₁₄H₁₇NO₃
MW: 247.2900 g/mol
MDL Number: MFCD05864717
CAS Number: 96551-22-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty