Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-piperidinobenzenecarbaldehyde ≥95%

Supplier: MATRIX SCIENTIFIC MS

043394-500MG 043394-1G 043394-5G
101860-560EA 278.46 USD InStock
101860-560 101860-562 101860-564
4-Bromo-2-piperidinobenzenecarbaldehyde ≥95%
4-Bromo-2-piperidinobenzenecarbaldehyde
Hazard: Irritant


Formula: C₁₂H₁₄BrNO
MDL Number: MFCD08276958
CAS Number: 643094-36-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty