Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-[5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]-1-ethanol ≥95%

Supplier: MATRIX SCIENTIFIC MS

043707-500MG 043707-1G 043707-5G
101862-158EA 556.92 USD InStock
101862-158 101862-160 101862-162
1-[5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]-1-ethanol ≥95%
1-[5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]-1-ethanol
Hazard: Irritant


Formula: C₇H₈ClF₃N₂O
MDL Number: MFCD04117790
CAS Number: 959578-12-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty