Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(4-Bromo-2-methylphenyl)-1H-pyrrole ≥95%

Supplier: MATRIX SCIENTIFIC MS

043994-500MG 043994-1G 043994-5G
101863-576EA 181.54 USD InStock
101863-576 101863-578 101863-580
1-(4-Bromo-2-methylphenyl)-1H-pyrrole ≥95%
1-(4-Bromo-2-methylphenyl)-1H-pyrrole
Hazard: Irritant


Formula: C₁₁H₁₀BrN
MW: 236.12 g/mol
MDL Number: MFCD02665256
CAS Number: 383137-70-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty