Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-Methoxyphenoxy)-1,3-thiazole-5-carbaldehyde ≥95%

Supplier: MATRIX SCIENTIFIC MS

045639-500MG 045639-1G 045639-5G
101871-878EA 921.54 USD InStock
101871-878 101871-880 101871-882
2-(4-Methoxyphenoxy)-1,3-thiazole-5-carbaldehyde ≥95%
2-(4-Methoxyphenoxy)-1,3-thiazole-5-carbaldehyde
Hazard: Irritant


Formula: C₁₁H₉NO₃S
MW: 235.26 g/mol
MDL Number: MFCD01871705
CAS Number: 478081-25-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty