Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Carbidopa monohydrate ≥98%

Supplier: MATRIX SCIENTIFIC MS

047882-1G 047882-5G 047882-25G
101881-836EA 44.62 USD InStock
101881-836 101881-838 102551-380
Carbidopa monohydrate ≥98%
Carbidopa monohydrate
Hazard: Irritant


Formula: C₁₀H₁₄N₂O₄
MW: 226.23 g/mol
MDL Number: MFCD00889211
CAS Number: 38821-49-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty