Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-3-ethyl-2,4(1H,3H)-quinazolinedione ≥95%

Supplier: MATRIX SCIENTIFIC MS

047963-500MG 047963-1G
101882-186EA 210.77 USD InStock
101882-186 101882-188
6-Bromo-3-ethyl-2,4(1H,3H)-quinazolinedione ≥95%
6-Bromo-3-ethyl-2,4(1H,3H)-quinazolinedione
Hazard: Irritant


Formula: C₁₀H₉BrN₂O₂
Melting Pt: 218…219 °C
MDL Number: MFCD00262937
CAS Number: 377067-64-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty