Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-Methyl-2,6-dinitroanilino)propanoic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

048123-500MG 048123-1G 048123-5G
101882-908EA 747.69 USD InStock
101882-908 101882-910 101882-912
2-(4-Methyl-2,6-dinitroanilino)propanoic acid ≥95%
2-(4-Methyl-2,6-dinitroanilino)propanoic acid
Hazard: Irritant


Formula: C₁₀H₁₁N₃O₆
MDL Number: MFCD05663836
CAS Number: 19466-60-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty