Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

cis-Methyl-4-hydroxy-2-piperidinecarboxylate ≥95%

Supplier: MATRIX SCIENTIFIC MS

048159-500MG 048159-1G
101883-090EA 710.77 USD InStock
101883-090 101883-092
cis-Methyl-4-hydroxy-2-piperidinecarboxylate ≥95%
cis-Methyl-4-hydroxy-2-piperidinecarboxylate
Hazard: Irritant


Formula: C₇H₁₃NO₃
MDL Number: MFCD09152760
CAS Number: 144913-66-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty