Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4-methoxyphenyl-3-pyrrolidinyl ether hydrochloride

Supplier: MATRIX SCIENTIFIC MS

050476-500MG 050476-2.5G
101891-804EA 364.62 USD InStock
101891-804 101891-806
2-Chloro-4-methoxyphenyl-3-pyrrolidinyl ether hydrochloride
2-Chloro-4-methoxyphenyl-3-pyrrolidinyl ether hydrochloride
Hazard: Irritant


Formula: C₁₁H₁₅Cl₂NO₂
MDL Number: MFCD13560877
CAS Number: 1220031-02-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty