Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-{[4-(1,1,3,3-Tetramethylbutyl)phenoxy]methyl}piperidine hydrochloride

Supplier: MATRIX SCIENTIFIC MS

050758-500MG 050758-2.5G
101892-928EA 364.62 USD InStock
101892-928 101892-930
4-{[4-(1,1,3,3-Tetramethylbutyl)phenoxy]methyl}piperidine hydrochloride
4-{[4-(1,1,3,3-Tetramethylbutyl)phenoxy]methyl}piperidine hydrochloride
Hazard: Irritant


Formula: C₂₀H₃₄ClNO
MDL Number: MFCD13561145
CAS Number: 1220034-04-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty