Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-6-(2,3-dihydro-1H-indol-1-yl)-1,3,5-triazin-2-amine

Supplier: MATRIX SCIENTIFIC MS

052098-500MG 052098-2.5G
101898-282EA 364.62 USD InStock
101898-282 101898-284
4-Chloro-6-(2,3-dihydro-1H-indol-1-yl)-1,3,5-triazin-2-amine
4-Chloro-6-(2,3-dihydro-1H-indol-1-yl)-1,3,5-triazin-2-amine
Hazard: Irritant


Formula: C₁₁H₁₀ClN₅
MDL Number: MFCD13561905
CAS Number: 1219967-86-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty