Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(6-Chloro-2-pyridinyl)-2-piperazinone

Supplier: MATRIX SCIENTIFIC MS

052176-500MG 052176-2.5G
101898-594EA 166.15 USD InStock
101898-594 101898-596
4-(6-Chloro-2-pyridinyl)-2-piperazinone
4-(6-Chloro-2-pyridinyl)-2-piperazinone
Hazard: Irritant


Formula: C₉H₁₀ClN₃O
MDL Number: MFCD13561960
CAS Number: 1220017-61-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty