Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N-Diethyl-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide hydrochloride

Supplier: MATRIX SCIENTIFIC MS

052557-500MG 052557-2.5G
101900-114EA 364.62 USD InStock
101900-114 101900-116
N,N-Diethyl-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide hydrochloride
N,N-Diethyl-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide hydrochloride
Hazard: Irritant


Formula: C₁₁H₁₉ClN₄O
MDL Number: MFCD13562291
CAS Number: 1220017-83-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty