Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2R)-1-(tert-Butoxycarbonyl)tetrahydro-1H-pyrrole-2-carboxylic-2-methylbutanoic anhydride ≥95%

Supplier: MATRIX SCIENTIFIC MS

055457-500MG 055457-1G
101909-370EA 278.46 USD InStock
101909-370 101909-372
(2R)-1-(tert-Butoxycarbonyl)tetrahydro-1H-pyrrole-2-carboxylic-2-methylbutanoic anhydride ≥95%
(2R)-1-(tert-Butoxycarbonyl)tetrahydro-1H-pyrrole-2-carboxylic-2-methylbutanoic anhydride
Hazard: Irritant


Formula: C₁₅H₂₅NO₅
MDL Number: MFCD16622769
CAS Number: 1301760-11-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty